missing translation for 'onlineSavingsMsg'
Learn More

Thyroglobulin Monoclonal Antibody (TG60), Invitrogen™

Mouse Monoclonal Antibody

$746.91 - $1,850.60

Specifications

Antigen Thyroglobulin
Clone TG60
Concentration 2.0 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIMA546102
View Documents
Invitrogen™
MA546102
100 μg N/A
Only null left
 
01-671-546
View Documents
Invitrogen™
MA110593
500 μg N/A
Only null left
 
PIMA545989
View Documents
Invitrogen™
MA545989
1 mg N/A
Only null left
 
Description

Description

MA1-10593 detects Thyroglobulin, specifically both T3 and T4, from human samples. MA1-10593 has been successfully used in ELISA applications. The MA1-10593 immunogen is purified human Thyroglobulin.

The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.
Specifications

Specifications

Thyroglobulin
2.0 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor
TG
IgG1
Protein A
Antibody
TG60
ELISA
Unconjugated
Mouse
Human
P01266
7038
Purified human Thyroglobulin.
Primary
-20°C, Avoid Freeze/Thaw Cycles
TG
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'