Learn More
Invitrogen™ Thyroglobulin Monoclonal Antibody (TG60), Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA110593
 View more versions of this product
Catalog No. 01-671-546
Only null left
Explore available promotions
Description
Description
MA1-10593 detects Thyroglobulin, specifically both T3 and T4, from human samples. MA1-10593 has been successfully used in ELISA applications. The MA1-10593 immunogen is purified human Thyroglobulin.
The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.
Specifications
Specifications
Thyroglobulin
Monoclonal
2.0 mg/mL
PBS with no preservative; pH 7.4
P01266
TG
Purified human Thyroglobulin.
500 μg
Primary
Human
Antibody
IgG1
ELISA
TG60
Unconjugated
TG
AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor
Mouse
Protein A
RUO
7038
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions