missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein Indicator, 0.1% (w/v) in 90% (v/v) Isopropyl Alcohol, Ricca Chemical

Catalog No. p-4770439
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOD1530100 100 mL Natural Poly Bottle
RSOD15301A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RSOD1530100 Supplier Ricca Chemical Company Supplier No. RSOD1530100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 125-20-2 , 67-63-0 , 7732-18-5
Molecular Formula C28H30O4
Molecular Weight (g/mol) Mixture
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Odor Characteristic alcohol odor
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Colorless
Solubility Information Solubility in Water: Infinite
Quantity 100 mL
Percent Purity 0.1% W/V
Grade Indicator
Chemical Name or Material Thymolphthalein Indicator, 0.1% (w/v) in 90% (v/v) Isopropyl Alcohols