missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein, 0.05% in Ethanol, Certified, LabChem™

$57.68 - $95.46

Specifications

Chemical Name or Material Thymolphthalein
Name Note 0.05% in Ethanol
CAS Max % 90.4,5,4.55,0.05
CAS 67-63-0 , 67-56-1 , 125-20-2
Linear Formula C28H30O4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC260707
View Documents
LabChem
LC260707
125 mL
Each for $57.68
Only null left
 
LC260701
View Documents
LabChem
LC260701
500 mL
Each for $95.46
Only null left
 
Description

Description

Specifications

Specifications

Thymolphthalein
90.4,5,4.55,0.05
C28H30O4
Passes Test
Certified
Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
9.3 to 10.5
0.05% in Ethanol
67-63-0 , 67-56-1 , 125-20-2
Soluble in water
430.5
Poly Bottle
Colorless to Blue
Liquid
Videos
SDS
Documents

Documents