missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymolphthalein, 0.05% in Ethanol, Certified, LabChem™
Supplier: LabChem LC260707
Specifications
| Thymolphthalein | |
| 90.4,5,4.55,0.05 | |
| C28H30O4 | |
| Passes Test | |
| Certified | |
| Fume; Carbon monoxide; Carbon dioxide; May release flammable gases | |
| 9.3 to 10.5 | |
| Liquid |
| 0.05% in Ethanol | |
| 67-63-0,67-56-1,125-20-2 | |
| Soluble in water | |
| 430.5 | |
| Poly Bottle | |
| Colorless to Blue | |
| 125 mL |