missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymol Blue Indicator, 0.4% (w/v), Ricca Chemical
$93.23 - $217.10
Chemical Identifiers
| CAS | 62625-21-2 |
|---|---|
| Molecular Formula | C27H29NaO5S |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00151093 |
| InChI Key | OCMIKNBSIRSUPI-FRWNMSGJSA-M |
| PubChem CID | 23692293 |
| IUPAC Name | sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate |
| SMILES | [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
Chemical Identifiers
| 62625-21-2 | |
| Mixture | |
| OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
| sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate |
| C27H29NaO5S | |
| MFCD00151093 | |
| 23692293 | |
| [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
Specifications
| 62625-21-2,7732-18-5 | |
| 100.0,0.41 | |
| C27H29NaO5S | |
| OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
| sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate | |
| 23692293 | |
| Indicator | |
| Aqueous Solution | |
| 9 | |
| Liquid |
| 98.0,0.39 | |
| 0.4% (w/v) | |
| MFCD00151093 | |
| [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O | |
| Mixture | |
| 0.4% W/V | |
| Natural Poly Bottle | |
| Dark Green | |
| 120 mL | |
| Thymol Blue Indicator |