missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue Indicator, 0.04% (w/v), Ricca Chemical

$68.97 - $138.30

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) Mixture
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
IUPAC Name sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
8400-4
View Documents
Ricca Chemical Company
84004
120 mL Natural Poly Bottle
Each for $68.97
Only null left
 
8400-16
View Documents
Ricca Chemical Company
RI840016
500 mL Natural Poly Bottle
Each for $138.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

62625-21-2
Mixture
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
C27H29NaO5S
MFCD00151093
23692293
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

62625-21-2 , 7732-18-5
100.0,0.04
C27H29NaO5S
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
23692293
Indicator
Aqueous Solution
9
Liquid
98.0,0.04
0.04% (w/v)
MFCD00151093
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Mixture
0.04% W/V
Natural Poly Bottle
Dark Green
120 mL
Thymol Blue Indicator
Videos
SDS
Documents

Documents