missing translation for 'onlineSavingsMsg'
Learn More

Thymol Blue, 0.04% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue, Certified, LabChem™

Supplier:  LabChem LC260207

Catalog No. LC260207


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Thymol Blue
62625-21-2,7732-18-5
C27H29NaO5S
MFCD00151093
Passes Test
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
488.57
488.58
Poly Bottle
Amber
Liquid
0.04% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue
99.96,0.04
C27H29NaO5S
Soluble in water
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
23692293
Certified
Sulfur compounds; Carbon monoxide; Carbon dioxide
125 mL

Chemical Identifiers

62625-21-2
488.57
OCMIKNBSIRSUPI-FRWNMSGJSA-M
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
C27H29NaO5S
MFCD00151093
23692293
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions