missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Thrombin Protease
Description
- Supplied as lyophilized powder
- Enables the site-specific cleavage of fusion proteins containing an accessible thrombin recognition sequence
- Purified from bovine plasma; functionally free of other clotting factors, plasminogen, and plasmin
- Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose™ 4B, Glutathione Sepharose High Performance, or Glutathione Sepharose 4 Fast Flow
Specifications
Specifications
| Buffer | Elution Buffer: 50mM Tris HCl, 10mM reduced Glutathione pH 8, Cleavage Buffer: PBS: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4 pH 7.3 |
| Product Type | Thrombin Protease Fusion Protein |
| Quantity | 500U |
| For Use With (Application) | Site-specific separation of the GST tag from proteins expressed using pGEX-T vectors, one unit of enzyme cleaves 90% of 100μg of a test GST fusion protein when incubated in 1X PBS at 22°C for 16 hours |