missing translation for 'onlineSavingsMsg'
Learn More

Thrombin, High purity grade, MP Biomedicals™

Catalog No. ICN15416310
Change view
Click to view available options
Quantity:
10kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15416310 10kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15416310 Supplier MP Biomedicals Inc Supplier No. 0215416310

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9002-04-4

Specifications

Color White
Grade High Purity
Quantity 10kU
Product Type Thrombin
Form Powder