Learn More
Corning™ PYREX™ Three-Way Angle Connecting Adapter, Standard Taper Joint
Three-way connecting tube with side arm placed at 75° angle from lower joint
Supplier: Corning™ 900019
Alert:
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Description
The sidearm is at an angle approximately 75° from the lower joint. The lower end and sidearm are equipped with inner joints, and the upper end with an outer joint. All joints on each tube are of the same size.
Chemical Identifiers
| Borosilicate Glass (3.3) | |
| 75° | |
| Tube Adapter |
Specifications
| Borosilicate Glass (3.3) | |
| 10.5 cm | |
| 75° |
| 4.13 in. | |
| Two 19/22 Inner Bottom/Outer Top | |
| Tube Adapter |