missing translation for 'onlineSavingsMsg'
Learn More

Thorium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLTH22Y

 View more versions of this product

Catalog No. PLTH2-2Y


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Th1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Thorium Standard (Depleted)
7440-29-1,7697-37-2,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
AA, ICP
Certified Reference Material
[Th]
thorium
125 mL
CHEBI:33385
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLTH2-2Y, -2X    10,000    PLTH2-3Y, -3X
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
0.1,2.0,97.9
Th
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ZSLUVFAKFWKJRC-UHFFFAOYSA-N
2% HNO3
232.038
23960
Assurance

Chemical Identifiers

7440-29-1
232.038
23960
thorium
Th
ZSLUVFAKFWKJRC-UHFFFAOYSA-N
CHEBI:33385
[Th]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only