missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thorium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$271.50 - $454.74
Chemical Identifiers
| CAS | 7440-29-1 |
|---|---|
| Molecular Formula | Th |
| Molecular Weight (g/mol) | 232.038 |
| InChI Key | ZSLUVFAKFWKJRC-UHFFFAOYSA-N |
| PubChem CID | 23960 |
| ChEBI | CHEBI:33385 |
| IUPAC Name | thorium |
| SMILES | [Th] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLTH2-2Y
|
SPEX CertiPrep Group LLC
PLTH22Y |
125 mL | Triple-leached LDPE bottle |
Each for $271.50
|
|
||||
|
PLTH2-2X
|
SPEX CertiPrep Group LLC
PLTH22X |
500 mL | Triple-leached LDPE bottle |
Each for $454.74
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Th | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-29-1 | |
| 232.038 | |
| 23960 | |
| thorium |
| Th | |
| ZSLUVFAKFWKJRC-UHFFFAOYSA-N | |
| CHEBI:33385 | |
| [Th] |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| 0.1,2.0,97.9 | |
| Th | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| ZSLUVFAKFWKJRC-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 232.038 | |
| 23960 | |
| Assurance | |
| Thorium Standard (Depleted) |
| 7440-29-1,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| AA, ICP | |
| Certified Reference Material | |
| [Th] | |
| thorium | |
| 125 mL | |
| CHEBI:33385 | |
| Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLTH2-2Y, -2X 10,000 PLTH2-3Y, -3X |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only