missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC13101EA

Quantity 10g

 View more versions of this product

Catalog No. CC13101EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

88-13-1
128.145
YNVOMSDITJMNET-UHFFFAOYSA-N
6918
C1=CSC=C1C(=O)O
C5H4O2S
MFCD00005467
3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be
thiophene-3-carboxylic acid

Specifications

3-Thiophenecarboxylic acid
96.5
97%
C5H4O2S
MFCD00005467
YNVOMSDITJMNET-UHFFFAOYSA-N
thiophene-3-carboxylic acid
6918
97%
88-13-1
100.0
Amber Glass Bottle
10g
3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be
C1=CSC=C1C(=O)O
128.145
128.15
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents