missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine 98.0+%, TCI America™

Catalog No. t10075ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
T10075ML 5 mL
T100725ML 25 mL
2 options

Catalog No. T10075ML

Supplier: TCI America T10075ML

Only null left

Chemical Identifiers

CAS 123-90-0
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00005974
InChI Key BRNULMACUQOKMR-UHFFFAOYSA-N
Synonym thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
PubChem CID 67164
ChEBI CHEBI:36392
IUPAC Name thiomorpholine
SMILES C1CSCCN1

Specifications

Color Yellow
Boiling Point 169°C
Quantity 5 mL
UN Number 3267
Formula Weight 103.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Thiomorpholine