accessibility menu, dialog, popup

UserName

HClSHN

CAS RN 123-90-0

IUPAC Name: hydrogen thiomorpholine chloride
Synonyms: thiomorpholine
Molecular Weight (g/mol): 139.64
Molecular Formula: C4H10ClNS
InChi Key: QSJAHJYXDRUZMY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1CSCCN1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)