missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiomorpholine 98.0+%, TCI America™
Supplier: TCI America T100725ML
Specifications
| Thiomorpholine | |
| Yellow | |
| C4H9NS | |
| 25 mL | |
| thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine | |
| C1CSCCN1 | |
| 103.183 | |
| CHEBI:36392 | |
| ≥98.0% (GC) |
| 123-90-0 | |
| 169°C | |
| MFCD00005974 | |
| 3267 | |
| BRNULMACUQOKMR-UHFFFAOYSA-N | |
| thiomorpholine | |
| 67164 | |
| 103.18 | |
| Liquid |
Chemical Identifiers
| 123-90-0 | |
| 103.183 | |
| BRNULMACUQOKMR-UHFFFAOYSA-N | |
| 67164 | |
| thiomorpholine |
| C4H9NS | |
| MFCD00005974 | |
| thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine | |
| CHEBI:36392 | |
| C1CSCCN1 |
Safety and Handling
TSCA : No