missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine, 98%

Catalog No. p-7022953
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1595806 5 g
AAA1595814 25 g
AAA1595822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1595806 Supplier Thermo Scientific Chemicals Supplier No. A1595806
Only null left

CAS: 123-90-0 | C4H9NS | 103.183 g/mol

Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

Solubility
Miscible with waterMiscible with water and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and air.

Chemical Identifiers

CAS 123-90-0
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00005974
InChI Key BRNULMACUQOKMR-UHFFFAOYSA-N
Synonym thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
PubChem CID 67164
ChEBI CHEBI:36392
IUPAC Name thiomorpholine
SMILES C1CSCCN1

Specifications

Density 1.086
Boiling Point 175°C to 177°C
Flash Point 67°C (153°F)
Refractive Index 1.5384
Quantity 5 g
UN Number UN3267
Beilstein 102550
Sensitivity Air Sensitive
Merck Index 14,9300
Solubility Information Miscible with waterMiscible with water and organic solvents.
Formula Weight 103.19
Percent Purity 98%
Chemical Name or Material Thiomorpholine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 204-660-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only