missing translation for 'onlineSavingsMsg'
Learn More

Thioisonicotinamide 97.0+%, TCI America™

Supplier:  TCI America T076425G

 View more versions of this product

Catalog No. T076425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Thioisonicotinamide
200°C
MFCD00006437
thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio
C1=CN=CC=C1C(=S)N
138.188
138.19
Crystalline Powder
2196-13-6
C6H6N2S
25 g
KPIIGXWUNXGGCP-UHFFFAOYSA-N
pyridine-4-carbothioamide
2723788
≥97.0% (T)

Chemical Identifiers

2196-13-6
138.188
KPIIGXWUNXGGCP-UHFFFAOYSA-N
2723788
C1=CN=CC=C1C(=S)N
C6H6N2S
MFCD00006437
thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio
pyridine-4-carbothioamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : US4204000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions