missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioisonicotinamide 97.0+%, TCI America™
$35.41 - $65.24
Chemical Identifiers
| CAS | 2196-13-6 |
|---|---|
| Molecular Formula | C6H6N2S |
| Molecular Weight (g/mol) | 138.188 |
| MDL Number | MFCD00006437 |
| InChI Key | KPIIGXWUNXGGCP-UHFFFAOYSA-N |
| Synonym | thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio |
| PubChem CID | 2723788 |
| IUPAC Name | pyridine-4-carbothioamide |
| SMILES | C1=CN=CC=C1C(=S)N |
Chemical Identifiers
| 2196-13-6 | |
| 138.188 | |
| KPIIGXWUNXGGCP-UHFFFAOYSA-N | |
| 2723788 | |
| C1=CN=CC=C1C(=S)N |
| C6H6N2S | |
| MFCD00006437 | |
| thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio | |
| pyridine-4-carbothioamide |
Specifications
| 2196-13-6 | |
| C6H6N2S | |
| 5 g | |
| KPIIGXWUNXGGCP-UHFFFAOYSA-N | |
| pyridine-4-carbothioamide | |
| 2723788 | |
| ≥97.0% (T) | |
| Thioisonicotinamide |
| 200°C | |
| MFCD00006437 | |
| thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio | |
| C1=CN=CC=C1C(=S)N | |
| 138.188 | |
| 138.19 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : US4204000
TSCA : Yes