missing translation for 'onlineSavingsMsg'
Learn More

Thioisobutyramide, 95%

Catalog No. p-7055944
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6466303 1 g
AAH6466306 5 g
AAH6466314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6466303 Supplier Thermo Scientific Chemicals Supplier No. H6466303
Only null left

CAS: 13515-65-6 | C4H9NS | 103.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13515-65-6
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD07369538
InChI Key NPCLRBQYESMUPD-UHFFFAOYSA-N
Synonym thioisobutyramide, isobutylthioamide, isobutyrthioamide, 2-methylpropaneothioamide, 2-methylpropane-thioamide, 2-methyl-propanethioamide, acmc-209by0, propanethioamide,2-methyl, ksc519m8d, 2-methyl propanethioamide
PubChem CID 10909507
IUPAC Name 2-methylpropanethioamide
SMILES CC(C)C(=S)N

Specifications

Melting Point 45°C to 48°C
Linear Formula (CH3)2CHC(=S)NH2
Quantity 1 g
Beilstein 1098427
Formula Weight 103.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Thioisobutyramide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only