CAS RN 13515-65-6
IUPAC Name:
2-methylpropanethioamide
Synonyms:
2-methylpropanethioamide
Molecular Weight (g/mol):
103.18
Molecular Formula:
C4H9NS
InChi Key:
NPCLRBQYESMUPD-UHFFFAOYSA-N
SMILES:
CC(C)C(N)=S
1
–
1
of
1
results
1
Promotions Available
Thioisobutyramide, 95%
Thioisobutyramide, 95%
CAS: 13515-65-6 Molecular Formula: C4H9NS Molecular Weight (g/mol): 103.183 MDL Number: MFCD07369538 InChI Key: NPCLRBQYESMUPD-UHFFFAOYSA-N Synonym: thioisobutyramide,isobutylthioamide,isobutyrthioamide,2-methylpropaneothioamide,2-methylpropane-thioamide,2-methyl-propanethioamide,acmc-209by0,propanethioamide,2-methyl,ksc519m8d,2-methyl propanethioamide PubChem CID: 10909507 IUPAC Name: 2-methylpropanethioamide SMILES: CC(C)C(=S)N