missing translation for 'onlineSavingsMsg'
Learn More

Thioglycolic Acid Solution, ∼80 in Water, MilliporeSigma™ Supelco™

Thioglycolic acid is an organic colourless compound with thiol and carboxylic acid as functional group. It is mainly used to make depilatories. It is considered to be corrosive to metals and tissues.

$247.90 - $247.90

Chemical Identifiers

MDL Number MFCD00004876
Synonym Mercaptoacetic acid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111018917
View Documents
Merck Emd Millipore
886521L
1 L
Each for $247.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00004876
Mercaptoacetic acid

Specifications

68-11-1
130°C - Closed Cup
MFCD00004876
Mercaptoacetic acid
80% aq. soln.
1.265 g/mL (at 20°C)
HSCH2 COOH
1 L
92.12
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'