missing translation for 'onlineSavingsMsg'
Learn More

Thiocyanate Standard Solution (0.1N (N/10)/Certified), Fisher Chemical

Catalog No. st181
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ST181 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST181 Supplier Thermo Fisher Scientific Supplier No. ST18 1
Only null left

Standardized against silver nitrate at 25°C

Chemical Identifiers

CAS 333-20-0 , 64-02-8 , 7732-18-5
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym Potassium Thiocyanate Solutions
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Packaging Poly Bottle
Identification Pass Test
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Physical Form Liquid
Concentration 0.0995 to 0.1005 N
Color Colorless
Quantity 1 L
Grade Certified
Chemical Name or Material Thiocyanate Standard Solution, 0.1N (N/10)