missing translation for 'onlineSavingsMsg'
Learn More

Thiocyanate Standard, 10 ppm SCN-, Ricca Chemical

Catalog No. p-3252563
Change view
Click to view available options
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
83944 Natural Poly Bottle
839416 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 83944 Supplier Ricca Chemical Company Supplier No. 83944

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 333-20-0 , 7732-18-5
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.0017
CAS Max % 100.0, 0.0017
Equivalence 1 mL = 0.01 mg SCN
Chemical Name or Material Thiocyanate Standard