accessibility menu, dialog, popup

UserName

CAS RN 2231-57-4

IUPAC Name: 1,3-diaminothiourea
Synonyms: thiocarbohydrazide
Molecular Weight (g/mol): 106.15
Molecular Formula: CH6N4S
InChi Key: LJTFFORYSFGNCT-UHFFFAOYSA-N
SMILES: NNC(=S)NN
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)