missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioanisole 99.0+%, TCI America™
Supplier: TCI America T0191500ML
Specifications
| Thioanisole | |
| Yellow | |
| C7H8S | |
| 500 mL | |
| thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene | |
| CSC1=CC=CC=C1 | |
| 124.201 | |
| 124.20 | |
| Liquid |
| 100-68-5 | |
| 188°C | |
| MFCD00008559 | |
| 1993 | |
| HNKJADCVZUBCPG-UHFFFAOYSA-N | |
| methylsulfanylbenzene | |
| 7520 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 100-68-5 | |
| 124.201 | |
| HNKJADCVZUBCPG-UHFFFAOYSA-N | |
| 7520 | |
| CSC1=CC=CC=C1 |
| C7H8S | |
| MFCD00008559 | |
| thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene | |
| methylsulfanylbenzene |
Safety and Handling
EINECSNumber : (3)-1098
RTECSNumber : DA6200000
TSCA : Yes