missing translation for 'onlineSavingsMsg'
Learn More

Thioanisole 99.0+%, TCI America™

Catalog No. p-7136133
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T019125ML 25 mL
T0191100ML 100 mL
T0191500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T019125ML Supplier TCI America Supplier No. T019125ML

Chemical Identifiers

CAS 100-68-5
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.201
MDL Number MFCD00008559
InChI Key HNKJADCVZUBCPG-UHFFFAOYSA-N
Synonym thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene
PubChem CID 7520
IUPAC Name methylsulfanylbenzene
SMILES CSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 mL
UN Number 1993
Formula Weight 124.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Thioanisole
EINECSNumber (3)-1098
RTECSNumber DA6200000
TSCA Yes