missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide TS, 4% w/v, Reagents

Catalog No. 01333459
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333459 1 L
01333458 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333459 Supplier Labchem Inc Supplier No. CT0104001C
Only null left

Thioacetamide TS, 4% w/v, Meets USP specifications, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F) Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 62-55-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Thiocarbamoyl Hydrazide, Aminothiourea, Hydrazinecarbothioamide
IUPAC Name water
SMILES O

Specifications

Color Colorless to Slight Yellow
Boiling Point 100°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.02
Percent Purity 4%
Grade USP
Physical Form Liquid
Chemical Name or Material Thioacetamide TS, 4% w/v