missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine 98.0+%, TCI America™

Catalog No. T07935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07935G 5 g
T079325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07935G Supplier TCI America Supplier No. T07935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 504-78-9
Molecular Formula C3H8NS
Molecular Weight (g/mol) 90.16
MDL Number MFCD00005211
InChI Key OGYGFUAIIOPWQD-UHFFFAOYSA-O
Synonym thiazolidine, tetrahydrothiazole, 1-thia-3-azacyclopentane, thiazole, tetrahydro, unii-i320806bkw, ccris 4275, acmc-1ao0m, 4-27-00-00009 beilstein handbook reference
PubChem CID 10444
ChEBI CHEBI:50120
IUPAC Name 1,3-thiazolidin-3-ium
SMILES C1CSC[NH2+]1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 g
UN Number 1993
Formula Weight 89.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Thiazolidine
RTECSNumber XJ5123700
TSCA No