accessibility menu, dialog, popup

UserName

CAS RN 504-78-9

IUPAC Name: 1,3-thiazolidin-3-ium
Synonyms: thiazolidine
Molecular Weight (g/mol): 90.16
Molecular Formula: C3H8NS
InChi Key: OGYGFUAIIOPWQD-UHFFFAOYSA-O
SMILES: C1CSC[NH2+]1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)