missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol

Catalog No. p-7042734
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6357503 1 g
AAJ6357506 5 g
AAJ6357514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6357503 Supplier Thermo Scientific Chemicals Supplier No. J6357503
Only null left

The methyl-sulfonyl analogue of chloramphenicol | CAS: 15318-45-3 | C12H15Cl2NO5S | 356.214 g/mol

Thiamphenicol is used to treat chancroid in men and uncomplicated gonorrhea. It is used in studies of bacterial protein synthesis at the level of peptidyl transferase activity associated with the 23S rRNA of the 50S ribosomal subunit. It is used to study chloraniphenicol-thiamphenicol-resistance and the use of fluorinated analogs when resistance is encountered. Its main advantage over chloramphenicol is that it has never been associated with aplastic anaemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiamphenicol is used to treat chancroid in men and uncomplicated gonorrhea. It is used in studies of bacterial protein synthesis at the level of peptidyl transferase activity associated with the 23S rRNA of the 50S ribosomal subunit. It is used to study chloraniphenicol-thiamphenicol-resistance and the use of fluorinated analogs when resistance is encountered. Its main advantage over chloramphenicol is that it has never been associated with aplastic anaemia.

Solubility
Soluble in acetonitrile or DMF. Slightly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 15318-45-3
Molecular Formula C12H15Cl2NO5S
Molecular Weight (g/mol) 356.214
MDL Number MFCD00467983
InChI Key OTVAEFIXJLOWRX-NXEZZACHSA-N
Synonym thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol
PubChem CID 27200
ChEBI CHEBI:32215
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 2819542
Merck Index 14,9301
Solubility Information Soluble in acetonitrile or DMF; Slightly soluble in water
Formula Weight 356.22
Physical Form Powder
Chemical Name or Material Thiamphenicol
EINECSNumber 239-355-3
RTECSNumber AB6680000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only