missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamphenicol, 99.5%, MP Biomedicals™
$1052.88 - $1052.88
Chemical Identifiers
| CAS | 15318-45-3 |
|---|---|
| Molecular Formula | C12H15Cl2NO5S |
| Molecular Weight (g/mol) | 356.214 |
| InChI Key | OTVAEFIXJLOWRX-NXEZZACHSA-N |
| Synonym | D -threo-2, 2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-(methyl- sulfonyl)phenethyl]-acetamide |
| PubChem CID | 27200 |
| ChEBI | CHEBI:32215 |
| IUPAC Name | 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
| SMILES | CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15684625
|
MP Biomedicals Inc
0215684625 |
25 g |
Each for $1,052.88
|
|
|||||
Chemical Identifiers
| 15318-45-3 | |
| 356.214 | |
| D -threo-2, 2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-(methyl- sulfonyl)phenethyl]-acetamide | |
| CHEBI:32215 | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O |
| C12H15Cl2NO5S | |
| OTVAEFIXJLOWRX-NXEZZACHSA-N | |
| 27200 | |
| 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
Specifications
| Thiamphenicol | |
| 15318-45-3 | |
| 25 g | |
| C12H15Cl2NO5S | |
| OTVAEFIXJLOWRX-NXEZZACHSA-N | |
| 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide | |
| 27200 | |
| 356.2 |
| Free-flowing Powder | |
| White | |
| 0.995 | |
| D -threo-2, 2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-(methyl- sulfonyl)phenethyl]-acetamide | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O | |
| 356.214 | |
| CHEBI:32215 | |
| Thiamphenicol |