missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MuA Transposase

Catalog No. F750
Change view
Click to view available options
Volume:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume
F750 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F750 Supplier Thermo Scientific™ Supplier No. F750

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

MuA Transposase, Entranceposon (M1-CamR), Entranceposon (M1-KanR), 5x Reaction Buffer for MuA Transposase, Control Target DNA, NotI Miniprimer

Transposon-based tools for functional analysis of proteins.

The Mutation Generation System™ Kit (MGS™ Kit) is designed for rapid construction of insertion mutation libraries for any kind of DNA clones. The system employs the highly efficient transposition machinery of the bacteriophage Mu to generate a pool of 15 bp insertion mutants that can be utilized in a variety of functional analyses of the encoded proteins or regulatory DNA regions.

Thermo Scientific transposon products are based on the transposition machinery of the bacteriophage Mu. During the lytic phase of the phage's life cycle the machinery replicates its genome by transposing repeatedly inside the host genome. The Mu transposition reaction has been modified into an in vitro reaction catalyzed by a single enzyme - MuA Transposase. In this system, one in vitro reaction is capable of generating more than a million transposon insertion clones.

Features
The Mutation Generation System (MGS Kit) and Stop Generation System (STOP Kit) were developed for functional analysis of proteins. These new transposon tools enable the creation of saturated libraries of mutated proteins in a single reaction with less hands-on time than any other method. The location of the transposon insertion in each mutated clone can be mapped by either PCR or sequencing. With MGS and STOP kits, thousands of mutated clones are ready for expression studies in just 2 to 3 days.

The MGS Kit contains the complete set of reagents for transposon-based linker scanning mutagenesis of any target protein. The MGS Entranceposons are designed for making subtle changes in the structure of a target protein by inserting 15 bp in-frame linkers throughout the corresponding target gene. This in-frame insertion allows for conservation of downstream sequences.

The STOPKit Entranceposons contain translational stop codons in all three reading frames within the terminal portion of the transposon sequence. The proprietary modification of the Stop Generation System makes it possible to generate a saturated C-terminal deletion library from virtually any target protein with a maximum addition of three amino acids.

Highlights

  • Efficient - Create saturated insertion libraries for sequencing and protein analysis in a single reaction
  • Fast - Decrease hands-on time compared to conventional methods
  • Random - Eliminate target site preference or insertion hot-spot
Applications
The STOP Kit generates truncated proteins for functional assays of:
  • Enzymes
  • Receptors
  • Structural proteins etc.
    The MGS Kit generates random fifteen basepair in vitro insertions into any target DNA for:
  • Rapid generation of in-frame five amino acid insertion libraries of any protein for functional analyses
  • Rapid and random mutagenesis of cloned promoters and other regulatory DNA regions
  • Random insertion of a NotI restriction enzyme site into any target DNA clone
Includes
  • MuA Transposase (F-750):
  • MuA Transposase (0.22 μg/μL, 20 μL)
  • 5x Reaction Buffer
MuA Transposase concentrated (F-750C):
  • MuA Transposase concentrated (1.1 μg/μL, 20 μL)
  • 5x Reaction Buffer
  • MuA Storage Buffer
Stop Generation System (STOP Kit) (F-703):
  • MuA Transposase (0.22 μg/μL in MuA storage buffer)
  • Entranceposon (STOP-KanR) (100 ng/μL in TE buffer)
  • 5x Reaction Buffer for MuA Transposase
  • DMSO 100%
  • Control Target DNA (370 ng/μL in TE buffer)
  • MuEnd-2 Primer (25 μM in dd water)
  • SeqE Primer (10 μM in dd water)
  • SeqW Primer (10 μM in dd water)
Mutation Generation System (MGS Kit) (F-701):
  • MuA Transposase (0.22 μg/μL in MuA storage buffer)
  • Entranceposon (M1-CamR) (100 ng/μL in TE buffer)
  • Entranceposon (M1-KanR) (100 ng/μL in TE buffer)
  • 5x Reaction Buffer for MuA Transposase
  • Control Target DNA (370 ng/μL in TE buffer)

Rapid generation of in-frame five amino acid insertion libraries of any protein for functional analyses, rapid and random mutagenesis of cloned promoters and other regulatory DNA regions, random insertion of a NotI restriction enzyme site into any target DNA clone

Specifications

Concentration 0.22 μg/μL
Form Solution
Format Tube
Reaction Speed Fast
Product Type Transposon Products
Starting Material DNA
Volume 20 μL
Are there any stability problems arising from the fact that the whole Entranceposon is inserted into the target plasmid? Is the Entranceposon capable of further transposition inside host cells? How stable are the target plasmids that carry the Entranceposon?

The Entranceposonshave been designed so that the presence of the MuA Transposase enzyme is an absolute requirement for any transposition activity. The Entranceposons do not contain any genes from the bacteriophage Mu; only the DNA sequences from the right end of the Mu genome that are responsible for the transposase binding. However, the Entranceposons contain >50 bp inverted terminal repeats. To avoid instability resulting from homologous recombination between the repeats, the use of a recA mutant E. coli strain is recommended.

Is there any background problem in the bacteriophage Mu transposition system that is used in your Transposon Products?

No. The Entranceposons that come with the TGS and MGS Kits are non-replicating linear DNA molecules that are not maintained inside E. coli cells.

For Research Use Only. Not for use in diagnostic procedures.