missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Bio-Tite™ Urine Specimen Containers

Easy, stackable storage

$333.53 - $564.13

Specifications

Product Type Specimen Container
Closure Type Dual-threaded Cap
Material Virgin Polypropylene
Color Clear
For Use With (Application) For stackable storage
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Volume (English) Closure Color Sterility Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Volume (English) Closure Color Sterility Price Quantity  
13-711-72
View Documents
Thermo Scientific™
020147
90 mL 3 oz. Yellow Non-sterile
Case of 400 Each for $470.59
Only null left
 
13-711-64
View Documents
Thermo Scientific™
030006
60 mL 2 oz. Orange Sterile
Case of 500 Each for $564.13
Only null left
 
13-711-67
View Documents
Thermo Scientific™
070000
120 mL 4 oz. Orange Sterile
Case of 100 Each for $333.53
Only null left
 
Description

Description

Easy, stackable storage

  • Leak-resistant containers offer superior safety and handling
  • Clear, virgin polypropylene provides better view of samples and prevents container from discoloring or becoming brittle
  • Bio-Tite and dual-threaded caps provide a leak-resistant seal for harsh transport conditions and pneumatic tube systems
  • Sterilized by gamma irradiation, which eliminates potential contamination
  • Grips, located at the top and bottom of container, provide easy-grip and handling with or without latex gloves
  • Instructions printed in English and Spanish on the labels

Order Info

Available with either 48 or 53mm opening.

Specifications

Specifications

Specimen Container
Virgin Polypropylene
For stackable storage
Dual-threaded Cap
Clear
Videos
SDS
Documents

Documents