missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution, extended mass range

Catalog No. PI88340
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88340 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88340 Supplier Thermo Scientific™ Supplier No. 88340

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Obtain both positive and negative ionization calibration of Thermo Scientific™ Triple Stage Quadrupole instruments using the Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution, Extended Mass Range, which is a mixture of 14 highly pure, ionizable components (mass range: 69 m/z to 2800m/z).

The Thermo Scientific Pierce Triple Quadrupole Calibration Solution, Extended Mass Range, is a mixture of 14 highly pure, ionizable components (mass range: 69 m/z to 2800m/z) designed for both positive and negative ionization calibration of Thermo Scientific Triple Stage Quadrupole instruments.

  • Improved mass accuracy - extended mass range designed to improve mass accuracy when compared to existing calibration solutions
  • Positive and negative mode calibration - single formulation with extended mass ranges for both ionization calibration modes
  • Highly validated - high-quality, rigorously tested formulation with lot-specific certificates of analysis
  • Ready to use - reference standard may be directly loaded into a syringe and injected into the instrument
  • Stable - store at 2 to 8°C for 18 months
The Pierce Triple Quadrupole Calibration Solution is a ready-to-use formulation that has been optimized for both positive and negative ionization calibration of instruments. The extended range masses (69 m/z to 2800m/z) improve performance of the Thermo Scientific Quantiva™ and Endura™ Triple Quadrupole mass spectrometers. Additionally, the Triple Quadrupole Calibration Solution, Extended Mass Range, is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.
The Pierce Triple Quadrupole Calibration Solution, Extended Mass Range, formulation contains 14 compounds that are ionizable in both positive and negative modes for instrument calibration. The formulation has been specifically optimized for use with the TSQ Endura™ Series and TSQ Quantiva™ Series instruments. The calibration solution is also compatible with the TSQ Quantum™ Series#, TSQ Discovery™ Series#, TSQ Quantum Ultra™ Series#, TSQ Quantum Access™ Series#, and TSQ Vantage™ Series# instruments using the list of recommended masses.
Positive mode (m/Z): 69.0447, 142.1590, 322.0481, 622.0289, 922.0098
Negative mode (m/Z): 59.0128, 112.9845, 301.9970, 601.9779, 1033.9870
For the calibration procedure use:
TSQ Series Getting Started Guide (70111-97152)

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Calibration Solution
Product Line Pierce
Content And Storage 1 x 10 mL
Store in refrigerator (2–8°C).
Starting Material Peptides, Small Molecules
Final Product Type Peptides, Small Molecules
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 10 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.