missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Hot Start II High-Fidelity PCR Master Mixes

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

Supplier:  Thermo Scientific™ F566L

 View more versions of this product

Catalog No. F566L


Only null left

Includes: Phusion Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/μL)
  • 5x Phusion HF Buffer
  • 5x Phusion GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Phusion Green Hot Start II High–Fidelity DNA Polymerase
  • Phusion Hot Start II High–Fidelity DNA Polymerase (2U/Glc muL)
  • 5x Phusion Green HF Buffer
  • 5x Phusion Green GC Buffer
  • DMSO
  • 50mM MgCl2 solution
Both Phusion and Phusion Green Buffer provide 1.5 mM MgCl2 in the final 1x concentration

Description

Description

Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Highlights

  • Reaction set up at room temperature
  • No non-specific amplification and primer degradation during reaction set up
  • Zero-time reactivation due to unique hot start technology
  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15-30 s/kb)
  • Robust reactions, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gels

With Phusion Hot Start II High-Fidelity DNA Polymerases amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3'→5' exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols.

Phusion Green Hot Start II High-Fidelity PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phusion Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2. The buffer also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel.

Applications

  • High-fidelity PCR
  • High throughput
  • Amplification of difficult (GC-rich) templates
  • Template generation for sequencing
  • Multiplex PCR
  • Long-range PCR
  • Cloning
  • Mutagenesis
  • Microarray

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

Specifications

Chemical Identifiers

2X
Phusion Hot Start II High-Fidelity DNA Polymerase
Hot Start High-Fidelity PCR Master Mix
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
SuperMix or Master Mix
Medium
Fast
Dry Ice
52X
Blunt
20 kb or less

Specifications

2X
Medium
Fast
500 reactions
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Blunt
20 kb or less
• 2X Phusion Green Hot Start II High-Fidelity PCR Master Mix (provides 1.5 mM MgCl2 in the final 1X concentration)
• Nuclease-free water
• DMSO

Store at -20°C.
Phusion Hot Start II High-Fidelity DNA Polymerase
Hot Start High-Fidelity PCR Master Mix
Dry Ice
52X
500 Reactions
SuperMix or Master Mix
Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.