Learn More
Thermo Scientific™ Low Protein Binding Microcentrifuge Tubes
Minimize protein sample loss during sample manipulation, especially when there are extensive processing steps, by using Thermo Scientific Pierce Low Protein Binding Microcentrifuge Tubes.
Supplier: Thermo Scientific™ 88380
Description
Thermo Scientific Pierce Low Protein Binding Microcentrifuge Tubes (1.5 or 2 mL) help minimize protein sample loss during sample manipulation and are ideal in applications where processing steps are extensive.
Pierce Low Protein Binding Microcentrifuge Tubes help minimize sample loss in mass spectrometry and immunoprecipitation/co-immunoprecipitation applications. The tubes are conveniently packaged at 250 tubes per package to minimize contamination even when purchased in bulk.• Durable—use at speeds up to 20,000 RCF
• Compatible—suitable for IP and mass spectrometry applications
• Nuclease-free—certified RNase/DNase-free polypropylene tubes
Chemical Identifiers
| Low Protein Binding Tube | |
| Pierce | |
| Collection Tube |
Specifications
| Low Protein Binding Tube | |
| 2 mL | |
| 2500 |
| 2 mL | |
| Pierce | |
| Collection Tube |