Learn More
Thermo Scientific™ MagJET Separation Rack, 12 x 1.5 mL tube
The MagJET separation rack allows fast and efficient procedures for magnetic bead-based nucleic acid isolation and affinity purification workflows.
Supplier: Thermo Scientific™ MR02
Description
Thermo Scientific MagJET Separation Rack is a magnetic separation device intended for low-throughput magnetic bead manipulation procedures such as magnetic bead-based nucleic acid isolation. The MagJET Separation Rack contains a high-performance permanent neodymium magnet embedded in a plastic separation unit frame. In combination with MagJET Magnetic Beads the MagJET Separation Rack allows fast and efficient magnetic bead-based separation procedures.
Highlights
• Contains a powerful permanent neodymium magnet for instantaneous bead collection from the solution
• Specially designed for robust performance with different sample volumes
• Stable and tight magnetic bead capture on microcentrifuge tube wall
• Highly efficient nucleic acid purification in combination with Thermo Scientific MagJET magnetic-bead based kits
Applications
• Affinity purification using magnetic particles.
Includes
• MagJET Separation Rack unit
Specifications
| MagJET Separation Rack, 12 x 1.5mL tubes | |
| Magnetic Stand | |
| MagJET Separation Rack |
| Tube | |
| MagJET | |
| 12 x 1.5 mL |
For Research Use Only. Not for use in diagnostic procedures.