missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagJET Separation Rack, 2 x 1.5 mL tube

The MagJET separation rack allows fast and efficient procedures for magnetic bead-based nucleic acid isolation and affinity purification workflows.

Supplier:  Thermo Scientific™ MR01

Catalog No. FERMR01


Only null left
Explore available promotions

Description

Description

Thermo Scientific MagJET Separation Rack is a magnetic separation device intended for low-throughput magnetic bead manipulation procedures such as magnetic bead-based nucleic acid isolation. The MagJET Separation Rack contains a high-performance permanent neodymium magnet embedded in a plastic separation unit frame. In combination with MagJET Magnetic Beads the MagJET Separation Rack allows fast and efficient magnetic bead-based separation procedures.

Highlights
• Contains a powerful permanent neodymium magnet for instantaneous bead collection from the solution
• Specially designed for robust performance with different sample volumes
• Stable and tight magnetic bead capture on microcentrifuge tube wall
• Highly efficient nucleic acid purification in combination with Thermo Scientific MagJET magnetic-bead based kits

Applications
• Affinity purification using magnetic particles.

Includes
• MagJET Separation Rack unit

Specifications

Specifications

MagJET Separation Rack, 2 x 1.5mL tubes
Magnetic Stand
MagJET Separation Rack
Tube
MagJET
2 x 1.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.