missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagJET Plasmid DNA Kit

Catalog No. FERK2792
Change view
Click to view available options
Quantity:
4 x 96 Preps
96 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK2792 4 x 96 Preps
FERK2791 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK2792 Supplier Thermo Scientific™ Supplier No. K2792

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.

Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.

The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET Magnetic Beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification for low sample throughput users.

The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors. Purified plasmid DNA can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.

Highlights
High yields—up to 5 μg of plasmid DNA from 1 mL of bacterial culture
Pure—isolated DNA is immediately ready-to-use in downstream application such as sequencing, transcription, digestion with restriction enzymes or transfection of robust cell lines
Flexible—automation and manual processing protocols available
Repeatable results and robust performance

Applications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
• FastDigest or conventional restriction digestion
• Automated fluorescent and radioactive sequencing
• PCR
• In vitro transcription
• Transformation
• Transfection of robust cell lines

Specifications

Format Kit
Isolation Technology Magnetic Bead
Sample Type Bacteria
For Use With (Application) Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual)
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line MagJET
Product Type Plasmid DNA Kit
Quantity 4 x 96 Preps
Target Plasmid DNA
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Are MagJET and GeneJET plasmid purification kits compatible with EndA+ bacterial strains?

Yes, MagJET and GeneJET Plasmid DNA Purification Kits are suitable for plasmid DNA purification from EndA+ strains. Please refer to the specific kit protocol for details. GeneJET Plasmid Miniprep Kit requires Wash Solution I (Cat. No. R1611, not included in the kit) for EndA+ strains.

Which magnetic rack should I use to follow the manual protocol of MagJET kits?

Thermo Scientific offers MagJET Separation Rack of different formats for 1.5 mL tubes (Cat. Nos. MR01, MR02), 50 mL tubes (Cat. No. MR04), and 96-well plates (Cat. No. MR03). You may use other compatible magnetic racks available in the market as well.

Is a sample size available for MagJET kits?

No, we do not offer a sample size for MagJET kits.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

For Research Use Only. Not for use in diagnostic procedures.