missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminaris HiGreen qPCR Master Mix

Catalog No. FERK0994
Change view
Click to view available options
Sufficient For:
250 Reactions
500 Reactions
5000 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
FERK0994 5000 Reactions
FERK0992 500 Reactions
FERK0991 250 Reactions
FERK0993 250 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERK0994 Supplier Thermo Scientific™ Supplier No. K0994

Includes

Luminaris HiGreen qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; Nuclease-Free Water

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

Ready-to-use qPCR master mixes with premixed UDG optimized for SYBR Green chemistry using standard cycling protocol. Master mixes available with or without blue color.

  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix

Specifications

Concentration 2X
Content And Storage Contains:
• 4 x 12.5 mL Luminaris HiGreen qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 5000 x 20 μL reactions
• 2 x 30 mL Nuclease-Free Water

Store at -20°C.
Detection Method SYBR
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
Product Line Luminaris
Product Type qPCR Master Mix
Quantity 5,000 reactions
Sufficient For 5000 Reactions
For Use With (Application) Gene Expression
Fidelity (vs. Taq) 2 X
No. of Reactions 5000 Reactions
Sample Type DNA, dsDNA
Show More Show Less
Does the yellow dye from the Yellow Sample Buffer provided with Luminaris Color and DyNAmo Flash qPCR Master Mixes influence qPCR performance?

The addition of yellow dye from the Yellow Sample Buffer to qPCR reaction has no influence on the efficiency, specificity, sensitivity or Cq values. Using the Yellow Sample Buffer is optional, but the dye helps to see in which wells the sample has been already added.

What DNA-binding dye is present in Luminaris Color HiGreen qPCR Master Mix?

Luminaris Color HiGreen master mixes contain SYBR Green I double-stranded DNA binding dye.

Does dUTP in Luminaris qPCR master mix influence qPCR efficiency?

Luminaris qPCR master mixes have all dTTP substituted with dUTP. dUTP presence does not influence the reaction speed and efficiency of the master mix. 100% substitution of dTTP with dUTP facilitates an efficient clearance of the contamination from previous PCR reactions.

What enzyme is used in Luminaris qPCR Master Mixes?

Luminaris qPCR master mixes contain Hot Start Taq DNA Polymerase. This is a Taq DNA polymerase, which has been chemically modified by the addition of heat-labile groups to amino acid residues blocking enzyme activity. The Hot Start Taq DNA Polymerase has ultra low residual activity until thermally activated. Therefore reactions can be assembled at room temperature with no risk for the extension of non-specifically annealed primers or primer dimers and providing higher specificity for DNA amplification. The enzyme is activated by 10 minutes incubation at 95 degrees C.

Can I prepare a qPCR plate in advance and store it for later use when using Luminaris qPCR Master Mixes?

When using a hot start polymerase it is not critical to start reactions immediately after setup. The Hot Start Taq DNA Polymerase in Luminaris qPCR Master Mixes has ultra low residual activity until thermally activated. Therefore reactions can be assembled and stored at room temperature for up to 72 hours with no risk for the extension of non-specifically annealed primers or primer dimers.

For Research Use Only. Not for use in diagnostic procedures.