missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ C8 Reversed Phase HPLC Column, 5 μm, 4.6 mm x 250 mm

Catalog No. 71405254630
Change view
Click to view available options
Length (Metric):
250 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
71405254630 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71405254630 Supplier Thermo Scientific™ Supplier No. 71405254630

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carbon Load: 7%; Pore Size: 150 Å; Column Phase: Reversed Phase

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Order Info

Other column dimensions are also available. Please call Customer Service for more information.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Particle Size 5 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L7
Stationary Phase C8
Product Line Beta
Packing Material High Purity, Base-Deactivated Silica
Carbon Load 7%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less