Learn More
Therapak™ Combination Temperature Biological Substance Category B Shipping Systems
Certified for ground and air shipments of Biological Substance Category B specimens
$185.18 - $611.21
Specifications
| Quantity | 1 Ea. |
|---|---|
| Capacity | 12 Tubes (6 per side) |
Includes: Transport box; UN3373 Biological Substance, dry ice label, and orientation arrow labels; 95kPa canisters/bags; absorbent material; list of contents card; cushioning material; and packing instructions
| Catalog Number | Mfr. No. | Includes | Conditions | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Conditions | Price | Quantity & Availability | ||||
|
22130034
|
Therapak™
37811 |
2 Med 95kPa Canister; 2 Aqui-Pak | Frozen/Ambient |
Case of 2 Each for $185.18
|
|
||||
|
22130033
|
Therapak™
37810 |
2 Med 95kPa Canister; 2 Aqui-Pak | Frozen/Refigerated |
Each for $204.97
|
|
||||
|
22130424
|
Therapak™
56726 |
Med 95kPa Canister, Bag; 2 Aqui-Pak | Frozen/Ambient |
Case of 6 Each for $611.21
|
|
||||
Description
- DOT and IATA Packing Instruction 650 compliant for the shipment of frozen/refrigerated or frozen/ambient specimens in one package
- Suitable for overnight shipments
Specifications
| 1 Ea. | |
| 12 Tubes (6 per side) |