Learn More
Therapak™ Combination Temperature Biological Substance Category B Shipping Systems
Certified for ground and air shipments of Biological Substance Category B specimens
Supplier:  Therapak™ 37811
 View more versions of this product
Catalog No. 22130034
Only null left
Includes: Transport box; UN3373 Biological Substance, dry ice label, and orientation arrow labels; 95kPa canisters/bags; absorbent material; list of contents card; cushioning material; and packing instructions
Description
Description
  • DOT and IATA Packing Instruction 650 compliant for the shipment of frozen/refrigerated or frozen/ambient specimens in one package
  • Suitable for overnight shipments
Specifications
Chemical Identifiers
1 Ea.
12 Tubes (6 per side)
Specifications
1 Ea.
Two medium 95kPa canisters; 2 Aqui-Pak absorbents; Cushioning; biohazard, Biological Substance Category B, arrow labels, contents card, transport box;
Frozen/Ambient
2 Med 95kPa Canister; 2 Aqui-Pak
12 Tubes (6 per side)
Product Suggestions
Product Suggestions