Learn More
Therapak™ Combination Temperature Biological Substance Category B Shipping Systems
Certified for ground and air shipments of Biological Substance Category B specimens
Supplier: Therapak™ 56726
Includes: Transport box; UN3373 Biological Substance, dry ice label, and orientation arrow labels; 95kPa canisters/bags; absorbent material; list of contents card; cushioning material; and packing instructions
Description
- DOT and IATA Packing Instruction 650 compliant for the shipment of frozen/refrigerated or frozen/ambient specimens in one package
- Suitable for overnight shipments
Chemical Identifiers
| 1 Ea. | |
| 12 Tubes (6 per side) |
Specifications
| 1 Ea. | |
| One medium 95kPa canister, bag; 2 Aqui-Pak absorbents, cushioning biohazard, Biological Substance Category B, arrow labels, list of contents card, transport box | |
| Frozen/Ambient |
| Med 95kPa Canister, Bag; 2 Aqui-Pak | |
| 12 Tubes (6 per side) |