missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC4-amine
Description
Thalidomide 4'-ether-alkylC4-amine is a functionalized cereblon ligand for PROTAC research and development; incorporates an E3 ligase ligand plus an alkyl linker with a terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 9
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-(4-Aminobutoxy)-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione hydrochloride |
| CAS | 2376990-29-1 |
| Synonym | Thalidomide - linker 9 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C17H19N3O5.HCl |
| Formula Weight | 381.81 |
| Product Type | Ligand |
| Purity | 95% |