Learn More
Texwipe™ AlphaMop™ Replacement Heads
AlphaMop™ 15 x 8 in. (38 x 20 cm) Thermoplastic Mop Head
Supplier:  Texwipe™ TX7108H
 View more versions of this product
Catalog No. 19-222-367
Only null left
Explore available promotions
Description
Description
  • The large mop head allows for wider surface coverage and faster cleaning results
  • Quick-change system allows the user to replace covers / pads frequently, adhering to strict cleaning protocols
  • The polyester pad assures the mop cover to conform to surfaces for consistent cleaning
  • Specifications
    Specifications
    Replacement Head
    Not Autoclavable
    No
    Non-sterile
    AlphaMop
    Thermoplastic
    Product Suggestions
    Product Suggestions