missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrapropylammonium Iodide 98.0+%, TCI America™
Supplier: TCI America T017225G
Specifications
| Tetrapropylammonium Iodide | |
| 285°C | |
| C12H28IN | |
| 25 g | |
| GKXDJYKZFZVASJ-UHFFFAOYSA-M | |
| tetrapropylazanium;iodide | |
| 12429 | |
| ≥98.0% (T) |
| 631-40-3 | |
| White | |
| MFCD00011842 | |
| tetrapropylammonium iodide, tetra-n-propylammonium iodide, ammonium, tetrapropyl-, iodide, tetrapropyl ammonium iodide, tetrapropylazanium iodide, n,n,n-tripropyl-1-propanaminium iodide, 1-propanaminium, n,n,n-tripropyl-, iodide, 1-propanaminium, n,n,n-tripropyl-, iodide 1:1, tetrapropylammoniumiodide, pr4ni | |
| CCC[N+](CCC)(CCC)CCC.[I-] | |
| 313.267 | |
| 313.27 | |
| Crystalline Powder |
Chemical Identifiers
| 631-40-3 | |
| 313.267 | |
| GKXDJYKZFZVASJ-UHFFFAOYSA-M | |
| 12429 | |
| CCC[N+](CCC)(CCC)CCC.[I-] |
| C12H28IN | |
| MFCD00011842 | |
| tetrapropylammonium iodide, tetra-n-propylammonium iodide, ammonium, tetrapropyl-, iodide, tetrapropyl ammonium iodide, tetrapropylazanium iodide, n,n,n-tripropyl-1-propanaminium iodide, 1-propanaminium, n,n,n-tripropyl-, iodide, 1-propanaminium, n,n,n-tripropyl-, iodide 1:1, tetrapropylammoniumiodide, pr4ni | |
| tetrapropylazanium;iodide |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : BS8410000
TSCA : Yes