missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrapropylammonium Iodide 98.0+%, TCI America™
$158.44 - $158.44
Chemical Identifiers
| CAS | 631-40-3 |
|---|---|
| Molecular Formula | C12H28IN |
| Molecular Weight (g/mol) | 313.267 |
| MDL Number | MFCD00011842 |
| InChI Key | GKXDJYKZFZVASJ-UHFFFAOYSA-M |
| Synonym | tetrapropylammonium iodide, tetra-n-propylammonium iodide, ammonium, tetrapropyl-, iodide, tetrapropyl ammonium iodide, tetrapropylazanium iodide, n,n,n-tripropyl-1-propanaminium iodide, 1-propanaminium, n,n,n-tripropyl-, iodide, 1-propanaminium, n,n,n-tripropyl-, iodide 1:1, tetrapropylammoniumiodide, pr4ni |
| PubChem CID | 12429 |
| IUPAC Name | tetrapropylazanium;iodide |
| SMILES | CCC[N+](CCC)(CCC)CCC.[I-] |
Chemical Identifiers
| 631-40-3 | |
| 313.267 | |
| GKXDJYKZFZVASJ-UHFFFAOYSA-M | |
| 12429 | |
| CCC[N+](CCC)(CCC)CCC.[I-] |
| C12H28IN | |
| MFCD00011842 | |
| tetrapropylammonium iodide, tetra-n-propylammonium iodide, ammonium, tetrapropyl-, iodide, tetrapropyl ammonium iodide, tetrapropylazanium iodide, n,n,n-tripropyl-1-propanaminium iodide, 1-propanaminium, n,n,n-tripropyl-, iodide, 1-propanaminium, n,n,n-tripropyl-, iodide 1:1, tetrapropylammoniumiodide, pr4ni | |
| tetrapropylazanium;iodide |
Specifications
| 631-40-3 | |
| White | |
| MFCD00011842 | |
| tetrapropylammonium iodide, tetra-n-propylammonium iodide, ammonium, tetrapropyl-, iodide, tetrapropyl ammonium iodide, tetrapropylazanium iodide, n,n,n-tripropyl-1-propanaminium iodide, 1-propanaminium, n,n,n-tripropyl-, iodide, 1-propanaminium, n,n,n-tripropyl-, iodide 1:1, tetrapropylammoniumiodide, pr4ni | |
| CCC[N+](CCC)(CCC)CCC.[I-] | |
| 313.267 | |
| 313.27 | |
| Crystalline Powder |
| 285°C | |
| C12H28IN | |
| 25 g | |
| GKXDJYKZFZVASJ-UHFFFAOYSA-M | |
| tetrapropylazanium;iodide | |
| 12429 | |
| ≥98.0% (T) | |
| Tetrapropylammonium Iodide |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : BS8410000
TSCA : Yes